EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Antibacterial activity and phytochemical analysis of Vochysia divergens (Vochysiaceae).
PMID 7500642
Abstract
Vochysia divergens Pohl (Vochysiaceae) is a tree commonly found in wet soils of 'Pantanal' of Mato Grosso, Brazil, and used in folk medicine against infections and asthma. We have studied different extracts and some isolated compounds from this plant for antibacterial activity. From the extracts of the stem bark beta-sitosterol, betulinic acid and sericic acid were isolated. The minimal inhibitory concentration (MIC) for Staphylococcus aureus were: ethanolic extract (MIC = 1.5 mg/ml); ethyl acetate extract (MIC = 2.0 mg/ml); and sericic acid (MIC = 1.0 mg/ml). Escherichia coli was resistant until 5 mg/ml.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00243
Sericic acid, ≥95% (LC/MS-ELSD)
C30H48O6