EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Halophilic protein stabilization by the mild solubilizing agents nondetergent sulfobetaines.
PMID 7503420
Abstract
In this work, experiments performed on pig heart and halophilic malate dehydrogenase as well as halophilic elongation factor Tu demonstrate a protein stabilization property from the recently described mild solubilizing agents nondetergent sulfobetaines. A practical application is given by the separation of halophilic bacteria elongation factor Tu and halophilic malate dehydrogenase by high-performance ion-exchange chromatography achieved at reduced salt levels without significant loss of activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N0665
NDSB 211, ≥98% (TLC)
C7H17NO4S