EMAIL THIS PAGE TO A FRIEND
Journal of immunological methods
Boc-Cys(Npys)-OH (BCNP): an appropriate reagent for the identification of T cell epitopes in cystine and/or cysteine-containing proteins.
PMID 7513735
Abstract
Some T cell epitopes become inactive when their thiols are blocked with various irreversible reagents (Régnier-Vigouroux, 1988; Maillère, 1992; Maillère et al., 1993). Blocking protein and peptide thiols with BCNP (Boc-Cys(Npys)-OH) constitutes a most appropriate strategy when searching for thiol-containing T cell epitopes. Free cysteines can thus be readily transformed into disulphide-like moieties which not only resist undesirable oxidative reactions but which also remain susceptible to reduction by antigen presenting cells, a prerequisite for the activity of thiol-dependent T cell epitopes. We describe the use of this reagent in a study of the intact disulphide-rich protein, toxin alpha from Naja nigricollis, and also two disulphide-containing toxin fragments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15554
Boc-Cys(Npys)-OH, ≥99.0% (TLC)
C13H17N3O6S2