EMAIL THIS PAGE TO A FRIEND
The EMBO journal
The decoding region of 16S RNA; a cross-linking study of the ribosomal A, P and E sites using tRNA derivatized at position 32 in the anticodon loop.
PMID 7516877
Abstract
A photo-reactive diazirine derivative was attached to the 2-thiocytidine residue at position 32 of tRNA(Arg)I from Escherichia coli. This modified tRNA was bound under suitable conditions to the A, P or E site of E.coli ribosomes. After photo-activation of the diazirine label, the sites of cross-linking to 16S rRNA were identified by our standard procedures. Each of the three tRNA binding sites showed a characteristic pattern of cross-linking. From tRNA at the A site, a major cross-link was observed to position 1378 of the 16S RNA, and a minor one to position 936. From the P site, there were major cross-links to positions 693 and to 957 and/or 966, as well as a minor cross-link to position 1338. The E site bound tRNA showed major cross-links to position 693 (identical to that from the P site) and to positions 1376/1378 (similar, but not identical, to the cross-link observed from the A site). Immunological analysis of the concomitantly cross-linked ribosomal proteins indicated that S7 was the major target of cross-linking from all three tRNA sites, with S11 as a minor product. The results are discussed in terms of the overall topography of the decoding region of the 30S ribosomal subunit.