EMAIL THIS PAGE TO A FRIEND
Blood
Increased fetal hemoglobin production in patients receiving valproic acid for epilepsy.
PMID 7520784
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
144436
Isobutyramide, 99%
C4H9NO