EMAIL THIS PAGE TO A FRIEND
International journal of pancreatology : official journal of the International Association of Pancreatology
Needle aspiration cytology in the diagnosis of mucinous cystadenocarcinoma of pancreas. A study of five cases with an emphasis on utility and differential diagnosis.
PMID 7520925
Abstract
In this study, our experience with five cases of mucinous cystadenocarcinoma of the pancreas is described, which was seen over a 13-yr period. The diagnosis in four of the cases was made from aspirated material that was obtained under computed tomography (CT) guidance and, in one case, following an intraoperative aspirate. In four cases, the tumor was found to be in the head of pancreas, whereas in one case, the neoplasm was located in the tail. All the cases were elderly male patients. As a result of our study, it is believed that aspiration cytodiagnosis of mucinous cystadenocarcinoma (a rare subtype of pancreatic carcinoma) can be made with confidence when performed intraoperatively or with imaging assistance. Also, aspiration cytology is a sensitive, specific, and relatively simple procedure that provides adequate material on which a confident diagnostic interpretation can be made of this uncommon neoplasm.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
41325 Mayer’s Mucicarmine Stain Solution, for microscopy