EMAIL THIS PAGE TO A FRIEND
In vitro cellular & developmental biology. Animal
Expression of some hepatocyte-like functional properties of WRL-68 cells in culture.
PMID 7522099
Abstract
Some morphologic and functional characteristics of an hepatic fetal human epithelial cell line (WRL-68 cells) were determined to validate the use of these cells as an in vitro hepatic model. WRL-68 cells have a morphologic structure similar to hepatocytes and hepatic primary cultures. They secrete alpha-feto protein and albumin and exhibit a cytokeratin pattern similar to other hepatic cultures. WRL-68 cells preserve the activity of some characteristic or specific liver enzymes or both used in clinical chemistry for the diagnosis of hepatic disorders, i.e. alanine amino transferase, aspartate amino transferase, gamma-glutamyl transpeptidase, and alkaline phosphatase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
WRL 68 (HeLa derivative), 89121403