EMAIL THIS PAGE TO A FRIEND
The Embo Journal
Association of the amino-terminal half of c-Src with focal adhesions alters their properties and is regulated by phosphorylation of tyrosine 527
PMID 7525268