EMAIL THIS PAGE TO A FRIEND
Journal of enzyme inhibition
Regulation of arylsulfate sulfotransferase from a human intestinal bacterium by nucleotides and magnesium ion.
PMID 7542320
Abstract
Arylsulfate sulfotransferase (ASST) from a human intestinal bacterium stoichiometrically catalyzed the transfer of a sulfate group from phenylsulfate esters to phenolic compounds. Pentachlorophenol, one of the selective inhibitors of phenol sulfoconjugation in mammalian tissues, inhibited both phenol and tyramine sulfation by ASST. Nucleotide triphosphates such as ATP, GTP, UTP and CTP, and pyrophosphate inhibited the ASST activity, whereas Mg2+ and Mn2+ activated the enzyme and prevented its inhibition by ATP and pyrophosphate. Equimolar binding of [alpha-] and [gamma-32P]ATP to the enzyme showed that the enzyme protein was not phosphorylated, but bound ATP. These results suggest that nucleotide triphosphates and divalent cations are important modulators in the control of ASST activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3877
Potassium 4-nitrophenyl sulfate, sulfatase substrate
C6H4KNO6S