EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Stimulation by 6-azauridine of de novo pyrimidine biosynthesis in BHK 165-23 cells.
PMID 7544952
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1882
6-Azauridine
C8H11N3O6