EMAIL THIS PAGE TO A FRIEND
Chromosome research : an international journal on the molecular, supramolecular and evolutionary aspects of chromosome biology
Imaging of DNA in human and plant chromosomes by high-resolution scanning electron microscopy.
PMID 7551552
Abstract
We describe a DNA-specific staining procedure, using a blue platinum organic dye, which allows DNA imaging of chromosomes by detection of back-scattered electrons in the scanning electron microscope. DNA distribution is visualized within chromosomal details of the centromeric and satellite regions, or in interphase chromatin, with high-resolution (10-15 nm) for comparison with the corresponding secondary electron image representing DNA plus protein.