EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Studies on the Chinese crude drug "shoma." IX. Three novel cyclolanostanol xylosides, cimicifugosides H-1, H-2 and H-5, from cimicifuga rhizome.
PMID 7553963
Abstract
Three new cyclolanostanol xylosides were isolated from a batch of commercial Cimicifuga Rhizome, cimicifugoside H-1 (1), C35H52O9, mp 260-262 degrees C, [alpha]D -43.5 degrees, cimicifugoside H-2 (2), C35H54O10, mp 227-229 degrees C, [alpha]D -38.8 degrees, and cimicifugoside H-5 (3), C35H52O10, mp 262-264 degrees C, [alpha]D -22.9 degrees, together with known glycosides, actein and 27-deoxyactein. Their structures were determined on the basis of chemical and spectrometric evidence including an X-ray crystallographic analysis. The structure of cimicifugoside H-1 (1) was established as (20R,24R)-24,25-epoxy-11 beta-hydroxy-3-beta-(beta-D- xylopyranosyloxy)-9,19-cyclolanost-7-ene-16,23-dione. Cimicifugoside H-5 (3) is the 15-hydroxylated derivative of 1. Since 1 changed into cimicifugoside H-2 (2) on treatment with p-toluenesulfonic acid, 2 has a 24R,25-diol structure derived from 1 by opening its epoxy ring.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1012032
Actein, United States Pharmacopeia (USP) Reference Standard
C37H56O11