EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Effects of chlorocresol (4-chloro-2-methyl phenol) administered during the fertilization and cleavage phases of Xenopus laevis.
PMID 7579923