EMAIL THIS PAGE TO A FRIEND
Archivio italiano di urologia, andrologia : organo ufficiale [di] Societa italiana di ecografia urologica e nefrologica
[Multicenter clinical study of the effects of once daily administration of mepartricin 150.000 U. in benign prostatic hypertrophy].
PMID 7581521
Abstract
The effects of partricin methyl ester, administered to 481 BPH patients at the dose of 150.000 U as a single administration at night for 90 days were studied in the course of a multicentre study. The favourable evolution of the objective and subjective symptomatologic parameters taken into consideration confirmed the efficacy of the substance used even by this mode of administration. The size of the patient sample ensured that reliable findings were obtained as regards the excellent safety of this form of dosage.