EMAIL THIS PAGE TO A FRIEND
Food additives and contaminants
Effects of some insect growth regulators on growth of Aspergillus flavus and its productivity of aflatoxin B1 and lipids.
PMID 7589721
Abstract
Two insect growth regulators, namely CME 134 and Dowco 439, have been tested for their effect on growth and on aflatoxin B1 and lipid productivity in Aspergillus flavus. No effect on growth and lipid production was detected. However, various concentrations of CME 134 and Dowco 439 showed significant inhibitory effects on aflatoxin B1 production. The degree of inhibition was dependent on the concentration and time of application. Inhibition was observed when Dowco 439 was added at inoculation and again 3, 6, 9, 12 and 15 days after inoculation, as well as after the addition of either CME 134 or Dowco 439 to the individual cultures at inoculation and 3 days after inoculation. However, the addition of either CME 134 or Dowco 439 as late as 6 days after inoculation markedly decreased the inhibition of aflatoxin B1 production.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45756
Teflubenzuron, PESTANAL®, analytical standard
C14H6Cl2F4N2O2