EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Mass spectrometric analysis of chitin oligosaccharides produced by Rhizobium NodC protein in Escherichia coli.
PMID 7592395
Abstract
A system for studying the in vivo activity of Rhizobium NodC protein in Escherichia coli has been developed. Using thin-layer chromatography, high-performance liquid chromatography, and mass spectrometry, we show that in this system R. leguminosarum bv. viciae NodC protein directs the synthesis of chitinpentaose, chitintetraose, chitintriose, and two as yet unidentified modified chitin oligosaccharides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T2144
N,N′,N′′-Triacetylchitotriose, ≥93% (HPLC)
C24H41N3O16