EMAIL THIS PAGE TO A FRIEND
Toxicon : official journal of the International Society on Toxinology
Chemical modification of cationic groups in the polypeptide cardiac stimulant anthopleurin-A.
PMID 7597722
Abstract
Chemical modification studies have been carried out on the sea anemone polypeptide anthopleurin-A in order to clarify the role of Arg-14 in its cardiac stimulatory activity. Reaction with 1,2-cyclohexanedione at 37 degrees C produced a range of protein products, including some with amino group modifications. These side-reactions were eliminated by prior citraconylation of the amino groups, which, following reaction with cyclohexanedione, could be reversed under conditions which preserved the cyclohexanedione adduct. Citraconylation of the three amino groups, one from the N-terminus and two from Lys-37 and Lys-48, destroyed the cardiac stimulatory activity of the molecule, but this was fully recoverable upon reversal of this reaction. It appears that one or more of the amino groups is essential for activity. Anthopleurin-A contains only one arginine residue, and this was confirmed as the site of modification by cyclohexanedione by showing that the product was refractory to proteolysis by trypsin, which normally cleaves the molecule at this residue. The positive inotropic activity of the cyclohexanedione adduct on isolated guinea-pig atria was identical to that of unmodified anthopleurin-A, indicating that the side-chain of Arg-14 is not required for cardiotonic activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C101400
1,2-Cyclohexanedione, 97%
C6H8O2
A7475 Anthopleurin-A, >88% (HPLC)
C220H326N64O67S6
125318
Citraconic anhydride, 98%
C5H4O3