EMAIL THIS PAGE TO A FRIEND
Naunyn-Schmiedeberg's archives of pharmacology
Benzimidazolones and renzapride facilitate acetylcholine release from guinea-pig myenteric plexus via 5-HT4 receptors.
PMID 7609775
Abstract
The effects of the 5-HT4 receptor agonists BIMU 8, BIMU 1, renzapride and of the 5-HT1p receptor agonist 5-hydroxyindalpine on basal and electrically evoked outflow of tritium were studied in guinea-pig longitudinal muscle myenteric plexus preparations preincubated with [3H]choline. Muscle contractions were recorded simultaneously. BIMU 8 caused a calcium dependent and tetrodotoxin sensitive increase in basal [3H]outflow that was assumed to represent release of [3H]acetylcholine. In addition, BIMU 8 enhanced the release of [3H]acetylcholine and twitch contractions evoked by submaximal electrical stimulation. Ondansetron (1 mumol/l) did not change the effects of BIMU 8, but DAU 6285 and tropisetron (each 1 mumol/l) competitively antagonized the various facilitatory effects of BIMU 8 with pA2 values of 7.0-7.2 (DAU 6285) and 7.0-7.3 (tropisetron). The phosphodiesterase inhibitors IBMX and rolipram did not increase the effects of BIMU 8. BIMU 1 and renzapride also concentration-dependently increased basal release of acetylcholine, and release and contractions caused by submaximal stimulation. The effects of BIMU 1 and renzapride were competitively antagonized by 1 mumol/l tropisetron (pA2 6.6-7.1). The EC50 values for the increase in the evoked [3H]acetylcholine release and contractions were closely similar. 5-Hydroxyindalpine did not change basal release and slightly inhibited the evoked release of [3H]acetylcholine. Release of acetylcholine and contractions elicited by submaximal stimulation were strongly inhibited by (+)-tubocurarine which indicates that nicotine ganglionic transmission is involved in this kind of release. The results suggest that BIMU 8, BIMU 1 and renzapride stimulate 5-HT4 receptors at cholinergic interneurones and thereby facilitate nicotinic ganglionic transmission in the myenteric plexus.(ABSTRACT TRUNCATED AT 250 WORDS)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4063
BIMU8 hydrate, ≥98% (HPLC)
C19H26N4O2·HCl · xH2O