EMAIL THIS PAGE TO A FRIEND
BioTechniques
Non-grinding method of DNA isolation from human pathogenic filamentous fungi using xanthogenates.
PMID 7619488
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
254770
Potassium ethyl xanthogenate, 96%
C3H5KOS2