EMAIL THIS PAGE TO A FRIEND
Experimental brain research
Dynamic differentiation of GABAA-sensitive influences on orientation selectivity of complex cells in the cat striate cortex.
PMID 7621943
Abstract
The influence of GABAA receptors on orientation selectivity of cat complex cells was tested by iontophoresis of the GABAA receptor blockers bicuculline and N-methyl-bicuculline while stimulating with drifting sinusoidal gratings. Reduction of orientation tuning was markedly less than reported in previous studies that used drifting bars as visual stimuli. Only 3/31 cells lost orientation selectivity, with an average increase in bandwidth of 33%, as opposed to half the cells losing selectivity and a bandwidth increase for the remainder of 47% as reported previously. Infusion of GABAA blockers revealed a prominent stimulus onset transient response, lasting about 120 ms, that showed a broadening of orientation selectivity comparable to that found using drifting bars under similar circumstances. We believe that drifting gratings emphasize a steady-state response component that retains, in the presence of GABAA blockers, significant orientation selectivity. Because the onset transient is initially unselective for orientation, we suggest that the steady-state, orientation-selective response component develops from an alternate inhibitory mechanism, possibly mediated by GABAB receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B7686
1(S),9(R)-(−)-Bicuculline methchloride, ≥97% (HPLC), powder
C21H20ClNO6