EMAIL THIS PAGE TO A FRIEND
Parasitology research
In vivo metabolism of aminopyrine by the larvae of the helminth Heligmosomoides polygyrus.
PMID 7624287
Abstract
The in vivo N-dealkylation of [13C-2]-labeled aminopyrine by the L1-L2 larvae of Heligmosomoides polygyrus was demonstrated by the use of a sensitive gas chromatography-mass spectrometry method. This is the first evidence for the possible existence of a cytochrome P-450-dependent activity in helminths.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
116416
1-Benzylimidazole, 99%
C10H10N2