EMAIL THIS PAGE TO A FRIEND
Allergologia et immunopathologia
Odor-associated idiopathic anaphylaxis. A case report.
PMID 7631593
Abstract
A 44 year old woman is described who appears to have idiopathic anaphylaxis triggered by chemical odors. Her case and a general discussion of anaphylaxis are presented. The known causes of anaphylaxis and a discourse on idiopathic anaphylaxis are given. The treatment of idiopathic anaphylaxis is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32142
Pirbuterol acetate, VETRANAL, analytical standard
C12H20N2O3 · C2H4O2