EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Biomedical applications
High-performance liquid chromatography-thermospray mass spectrometry of omega-carboxyleukotriene B4 and omega-hydroxyleukotriene B4 from an incubation mixture of human colonic well-differentiated adenocarcinoma homogenate.
PMID 7633595
Abstract
A method for the analysis of omega-carboxyleukotriene B4 and omega-hydroxyleukotriene B4 in human colonic carcinoma homogenate is described. The hydroxy groups of the leukotriene metabolite were acetylated by acetic anhydride, and the mixture was partially purified on a Sep-Pak C18 cartridge and analysed by reversed-phase HPLC-thermospray MS. Generally, the base ion, [MH-2(60)]+, is produced through elimination of two acetic acid (60 mass units) molecules from the protonated molecular ion. On selected-ion monitoring, standard curves for omega-carboxy- or omega-hydroxyleukotriene B4 showed a linear relationship over the range 72-1500 pmol. The assay based on selected-ion monitoring was applied to an extract from human colonic carcinoma homogenate. When a homogenate of human colonic well-differentiated adenocarcinoma was incubated with NADPH and leukotriene B4 (60.6 nmol) as a substrate, the conversion of precursor leukotriene B4 to omega-carboxyleukotriene B4 or omega-hydroxyleukotriene B4 was 0.33 or 3.17%, respectively. Based on these results, it is suggested that carcinoma cells themselves or leukocytes at the hostsite in a region of human colonic well-differentiated adenocarcinoma are performing omega-oxidation through NADPH-dependent omega-hydroxylation of leukotriene B4.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6046
20-Carboxy-leukotriene B4, ~1 mg/mL in ethanol, ≥90% (HPLC)
C20H30O6