EMAIL THIS PAGE TO A FRIEND
Atencion primaria
[Mild organophosphate poisoning: a lesser evil?].
PMID 7647200
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33383
Triasulfuron, PESTANAL®, analytical standard
C14H16ClN5O5S