EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Enzymatic transformation of de-O-acetyl-lanatoside C into digoxin.
PMID 7648578
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0600000 Deslanoside, European Pharmacopoeia (EP) Reference Standard
C47H74O19