EMAIL THIS PAGE TO A FRIEND
Indian journal of physiology and pharmacology
Evaluation of antimuscarinic activity in human volunteers: a teaching aid in clinical pharmacology.
PMID 7649608
Abstract
The antimuscarinic activity of oxyphenonium bromide, diphenhydramine hydrochloride and astemizole were evaluated in six volunteers. The parameters used were salivary secretion, heart rate and pupillary size. The results indicated that the changes in heart rate and pupillary size and measurements were not convenient parameters for class room demonstration. However, salivary secretion and dryness of mouth were found to be reliable parameters for measurement. It was concluded that simple procedures like evaluation of antimuscarinic activity could be introduced as teaching aids in clinical pharmacology for undergraduate students.