EMAIL THIS PAGE TO A FRIEND
AIDS (London, England)
HIV-2-specific cytotoxic T-lymphocyte activity is inversely related to proviral load.
PMID 7662192
Abstract
To characterize HIV-specific cytotoxic T-lymphocyte (CTL) activities in HIV-2-infected individuals and to relate these to HIV-2 proviral load. Peripheral blood mononuclear cells were collected from 16 HIV-2-seropositive and four HIV-1/2 dually seropositive subjects. CTL were restimulated with autologous phytohaemagglutinin-stimulated blasts and CTL activities in 'bulk' cultures were evaluated 7 and 14 days later by a standard 51Cr-release assay using autologous B-cell lines infected with recombinant vaccinia expressing HIV-2 Gag, Pol or Nef protein. Proviral load was quantified by polymerase chain reaction (PCR) which used HIV-2 long terminal repeat primers and an external standard control made by an HIV-2CBL-22 chronically infected C8166 cell line. A biotinylated primer was used to capture the 35S dATP-incorporated secondary PCR product in a quantitative radiometric assay. After 14 days of culture CTL responses against Gag or Pol protein were seen in 18 (90.0%) and 14 (70.0%) out of 20 subjects, respectively, whereas a CTL response was noted against Nef protein in five (25.0%) out of 20 subjects. In 14 (70.0%) out of 20 subjects multiple HIV proteins were simultaneously recognized. The sum of specific lysis (%) against HIV-2 Gag, Pol and Nef at 30:1 effector-to-target ratio, or specific lysis of the dominant CTL response, correlated strongly with HIV-2 proviral load expressed as copies per 10(5) CD4+ cells (r = -0.625, P = 0.003 and r = -0.674, P = 0.001, respectively). HIV-2-specific CTL to multiple gene products was demonstrated in most HIV-2-infected individuals. An inverse correlation between the level of CTL activity and proviral load was found, which supports the hypothesis that CTL are important in the control of HIV-2 replication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
88051601
C8166