EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Unique cholesteryl glucosides in Helicobacter pylori: composition and structural analysis.
PMID 7665522
Abstract
A chloroform-methanol-extracted lipid of Helicobacter pylori was studied. Three kinds of glycolipids, accounting for about 25% (wt/wt) of the total lipid, were detected and identified to be cholesteryl glucosides. The structures of two of them were determined to be cholesteryl-alpha-D-glucopyranoside and cholesteryl-6-O-tetrade-canoyl-alpha-D-glucopyranoside, and the plausible structure of the third one was identified as cholesteryl-6-O-phosphatidyl-alpha-D-glucopyranoside. Cholesteryl glucosides are very rare in animals and bacteria. Furthermore, those in H. pylori had an alpha-glycosidic linkage, which is rather unusual for natural glycosides, and a phosphate-linked cholesteryl glycoside like the cholesteryl-6-O-phosphatidyl-alpha-D-glucopyranoside has not been reported previously. As the cholesterol glucosides were detected in strains obtained from diverse geographical locations, the presence of cholesteryl glucosides in H. pylori is a very unique and a characteristic feature of the species. These findings add a new facet to the physiology and biochemistry, especially the cholesterol and glucose metabolism, of H. pylori. Furthermore, the cholesteryl glucosides of H. pylori showed hemolytic activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
97617
Cholesterol α-D-glucoside, ≥98% (TLC)
C33H56O6
28609
Cholesterol β-D-glucoside, ≥97% (TLC)
C33H56O6