EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
Inhibition of citrate oxidation in vitro by 2-hydroxybutyrate and 4-hydroxybutyrate in cerebral cortex of young rats.
PMID 7672250
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54917
(R)-2-Hydroxybutyric acid, ≥97.0% (T)
C4H8O3