EMAIL THIS PAGE TO A FRIEND
Applied biochemistry and biotechnology
Fungal degradation of organophosphorus insecticides.
PMID 7686734
Abstract
Organophosphorous insecticides are used extensively in agriculture. As a group, they are easily degraded by bacteria in the environment. However, a number of them have half-lives of several months. Little is known about their biodegradation by fungi. We showed that Phanerochaete chrysosporium mineralized chlorpyrifos, fonofos, and terbufos (27.5, 12.2, and 26.6%, respectively) during an 18-d incubation in nutrient nitrogen-limited cultures. Results demonstrated that the chlorinated pyridinyl ring of chlorpyrifos and the phenyl ring of fonofos undergo cleavage during biodegradation by the fungus. The usefulness of P. chrysosporium for bioremediation is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45313
Terbufos, PESTANAL®, analytical standard
C9H21O2PS3