EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Photocrosslinking of 5-iodouracil-substituted RNA and DNA to proteins.
PMID 7694369
Abstract
5-Iodouracil-substituted RNA and DNA were crosslinked regiospecifically to associated proteins in yields of 70 to 94% of bound nucleic acid. Irradiation of the iodouracil chromophore with monochromatic, long-wavelength ultraviolet radiation (325 nanometers) eliminates excitation of other nucleic acid and protein chromophores. The combination of high crosslinking yields, excellent specificity, and elimination of photodamage to other chromophores represents an important advance toward the precise identification of contacts in nucleoprotein complexes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
857858
5-Iodouracil, 98%
C4H3IN2O2