EMAIL THIS PAGE TO A FRIEND
The Annals of pharmacotherapy
Twice-daily cefaclor therapy.
PMID 7696725
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6895
Cefaclor
C15H14ClN3O4S