EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
1L-2,3:4,5-bis-O-(tetraisopropyldisiloxane-1,3-diyl)-chiro-inositol: a useful intermediate for the preparation of several novel cyclitols.
PMID 7697660