EMAIL THIS PAGE TO A FRIEND
Klinicheskaia khirurgiia
[Antiviral action of the macromolecular derivative of the platinum with deoxyribonucleic acid in the treatment of patient with malignant liver neoplasm].
PMID 7707643
Abstract
Results of the action of a preparation consisting of cis-dichlorodiammineplatinum and deoxyribonucleic acid combined in the saline balanced solution by means of the original technique on cytomegalovirus infection of patients with cancer of the liver are presented. Clearly pronounced antivirus activity of the preparation developed is noted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
244929
Dichloro(ethylenediamine)platinum(II), 99%
C2H8Cl2N2Pt