EMAIL THIS PAGE TO A FRIEND
Journal of neural transmission. Parkinson's disease and dementia section
Pyritinol facilitates the recovery of cortical cholinergic deficits caused by nucleus basalis lesions.
PMID 7710671
Abstract
The effect of a nootropic, Pyritinol, on the recovery of cortical cholinergic deficits induced by injury of the nucleus basalis has been tested on two groups of unilateral quisqualic acid nbM-lesioned rats. The first group had a 30 nmol lesion producing a cortical cholinergic impairment at 21 days, with a spontaneous recovery at 45 days. The second group had a 50 nmol lesion that produced a deeper cholinergic deficit, which did not recover at 45 days. Pyritinol enhanced the recovery in the 30 nmol group of animals on the 21st day after surgery. The recovery was measured as an increase in the activities of acetylcholinesterase (AChE), choline acetyltransferase (ChAT) and the high affinity choline uptake system, and the histochemical densities of the cortical AChE network and the M2 receptor. Histochemical analysis of the nbM enabled cortical recovery to be related to the number of surviving neurons and also to their hypertrophy and AChE-ChAT hyperactivity. Pyritinol enhanced recovery in 30 nmol lesioned animals but in the other group, with a lower number of surviving neurons and a lower ability of the cells to become hypertrophic, the drug was unable to promote cortical recovery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7171
Pyrithioxin, analytical standard
C16H20N2O4S2
P5156
Pyrithioxin dihydrochloride, analytical standard
C16H20N2O4S2 · 2HCl