EMAIL THIS PAGE TO A FRIEND
Fiziolohichnyi zhurnal (Kiev, Ukraine : 1994)
[Protective effect of vitamin E in acute administration of nitrobenzene and its chlorine derivatives into the stomach of rats].
PMID 7713233
Abstract
The level of lipid peroxidation, content of vitamin E and its metabolites as well as antioxidative activity in the blood serum, liver and spleen of white rats were studied. Toxicological effects of nitrobenzene and nitrochlorbenzenes were decreased by vitamin E.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185760
1-Chloro-2-nitrobenzene, 99%
C6H4ClNO2