EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Multiple GABA receptor subtypes mediate inhibition of calcium influx at rat retinal bipolar cell terminals.
PMID 7722621
Abstract
Inhibitory effects of GABA on K(+)-evoked Ca2+ influx into rat retinal bipolar cell terminals were studied using calcium imaging methods. Application of high K+ evokes a sustained, reversible increase in [Ca2+]i at bipolar cell terminals, which occurs mainly via dihydropyridine-sensitive (L-type) Ca2+ channels. There are at least two GABA receptor subtypes coexisting at bipolar cell terminals: a conventional GABAA receptor and a bicuculline/baclofen-insensitive GABA receptor. Activation of either GABA receptor inhibited the K(+)-evoked Ca2+ response. However, these two GABA receptor subtypes have distinct properties. GABAA receptors suppress the Ca2+ response only at relatively high concentrations of agonist, and with fas kinetics and a narrow dynamic range. In contrast, the bicuculline/baclofen-insensitive GABA receptors produce inhibition on the Ca2+ response at a much lower concentration of agonist, and with slow onset and a wider dynamic range. The pharmacologic profile of the bicuculline/baclofen-insensitive GABA receptor at bipolar cell terminals is most similar to the GABAC receptor reported by Feigenspan et al. (1993). Unlike the GABAC receptors described in other species, it is extremely insensitive to picrotoxin. Therefore, it may be appropriate to refer to this receptor as a picrotoxin-insensitive GABAc receptor. 3-Aminopropyl(methyl)phosphinic acid (3-APMPA) and 3-aminopropylphosphonic (3-APA), two phosphate analogs of GABA, selectively antagonize the picrotoxin-insensitive GABAc receptors but not the GABAA receptors in this system. These results imply a functional role for multiple GABA receptors in regulating synaptic transmission at bipolar cell terminals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
268615
3-Aminopropylphosphonic acid, 98%
C3H10NO3P