EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Diazoxide.
PMID 772431
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0950000
Diazoxide, European Pharmacopoeia (EP) Reference Standard
C8H7ClN2O2S