EMAIL THIS PAGE TO A FRIEND
Revue de pneumologie clinique
[Interstitial pneumopathy in a woman treated with althiazide].
PMID 7724984
Abstract
The authors report the case of interstitial pneumopathy in a woman treated with althiazide, thiadizical diuretic. This observation bring the discussion on responsibility to this drug in the origin of this pulmonary disease.