EMAIL THIS PAGE TO A FRIEND
Archives of dermatological research
Genotraumatic T cells and cutaneous T-cell lymphoma. A causal relationship?
PMID 7726642
Abstract
Mycosis fungoides, or cutaneous T-cell lymphoma (CTCL), is a T-cell mediated chronic inflammatory skin disease, which can occasionally progress with a variable time course to a fatal lymphoma or to a leukaemic form called Sézary's syndrome. Extensive research into CTCL has not yet elucidated the primary pathophysiological mechanisms. Immunohistological studies are so far less helpful than expected in establishing early diagnosis and prognosis of the disease. The proposition that an exogenous virus is the cause of CTCL has not been substantiated. Karyotypic analysis of lymphocytes from the skin and blood of patients with CTCL have shown the existence of several genetically aberrant T-cell clones in the same patient. These changes are discussed as potential primary events for the development of CTCL. The hypothesis is put forward that the development of genotraumatic T lymphocytes is involved in the progression of the disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
My-La CD4+, 95051032
details
My-La CD8+, 95051033