EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Standardized method for high-resolution 1H-NMR of cerebrospinal fluid.
PMID 7729054
Abstract
This study describes a standardized method for recording single-pulse 1H-nuclear magnetic resonance (1H-NMR) spectra from cerebrospinal fluid (CSF). Quantitative data for alanine, valine, threonine, and lactic acid correlated well with data obtained with conventional techniques. The pH of the samples is important for the reproducibility of the chemical shift of resonances, and should be standardized to improve recognition and assignment of resonances. A database of resonances from various metabolites is presented. Fifty compounds could be identified in CSF, 15 of which had not been observed earlier in NMR studies of CSF. We describe for the first time in the literature, to our knowledge, 3-hydroxyisovaleric acid as a regular component of many CSF samples. As examples of the diagnostic power of the technique, spectra are shown of CSF from patients with three different inborn errors of metabolism. We found high concentrations of N-acetylaspartic acid, citric acid, and succinic acid in CSF from a patient with Canavan disease. This is indirect evidence for the existence of a carrier mechanism that is shared by these di- and tricarboxylic acids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54920
(R)-3-Hydroxybutyric acid, ≥98.0% (T)
C4H8O3
27109
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
C6H8O7
A5835
γ-Aminobutyric acid, BioXtra, ≥99%
C4H9NO2
A2129
γ-Aminobutyric acid, ≥99%
C4H9NO2
L1750
L-(+)-Lactic acid, ≥98%
C3H6O3
L6402
L-(+)-Lactic acid, BioXtra, ≥98% (titration)
C3H6O3
27715
L-(+)-Lactic acid, 80%
C3H6O3
46937
L-(+)-Lactic acid, analytical standard
C3H6O3
L1875
L-(+)-Lactic acid solution, 30% in H2O (by weight)
C3H6O3
27714
L-(+)-Lactic acid solution, meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
C3H6O3
69780
L-(+)-Lactic acid solution, ~40% in H2O
C3H6O3
44526
L-Alanine, certified reference material, TraceCERT®
C3H7NO2
PHR1110
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO2
G4501
L-Glutathione oxidized, ≥98%, lyophilized powder
C20H32N6O12S2
G4376
L-Glutathione oxidized, ≥98% (HPLC)
C20H32N6O12S2
G6654
L-Glutathione oxidized, BioXtra, ≥98%
C20H32N6O12S2
V0500
L-Valine, reagent grade, ≥98% (HPLC)
C5H11NO2
V0513
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C5H11NO2
V4638
L-Valine, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C5H11NO2
94619
L-Valine, BioUltra, ≥99.5% (NT)
C5H11NO2
50848
L-Valine, certified reference material, TraceCERT®
C5H11NO2
W527718
L-Valine, FCC
C5H11NO2
199257
L-(+)-Lactic acid solution, in H2O, ≥85%
C3H6O3
PHR1172
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
C5H11NO2
00920
N-Acetyl-L-aspartic acid, ≥99.0% (T)
C6H9NO5
A5930
N-Acetyl-Asp-Glu, ≥97% (TLC), powder
C11H16N2O8
72644
Pyruvic acid, technical, ≥95.0% (T)
C3H4O3
69612
Succinic acid, matrix substance for MALDI-MS, ≥99.5% (T), Ultra pure
C4H6O4
49893
Succinic acid, certified reference material, TraceCERT®
C4H6O4
14080
Succinic acid, purum p.a., ≥99.0% (T)
C4H6O4