EMAIL THIS PAGE TO A FRIEND
The Biochemical Journal
Nerve growth factor stimulates a novel protein kinase in PC-12 cells that phosphorylates and activates mitogen-activated protein kinase kinase (MEK).
PMID 7733891