EMAIL THIS PAGE TO A FRIEND
Melanoma research
Tetramodality treatment of human melanoma in vitro.
PMID 7734956
Abstract
We evaluated the in vitro cytotoxic effects of combined human tumour necrosis factor alpha (TNF), human interferon gamma (IFN-gamma), melphalan (L-PAM) and hyperthermia (HTX) on human melanoma cell lines using the crystal violet assay. HTX (40 degrees C, 1 h) alone had no effect. The responses of the cell lines to TNF were in the rank order of 939 cells > 987 > 284 > C8161 > 852 > A2058 approximately 0, and all displayed shallow dose-response curves; no significant thermal enhancement of TNF cytotoxicity was apparent with this heat dose. All cell lines were sensitive to L-PAM, with 284 cells being the most sensitive; HTX caused only slightly increased sensitization to L-PAM. The combination of TNF and L-PAM resulted in generally subadditive or additive cytotoxicity, with or without HTX. The response to IFN-gamma alone was heterogeneous; the 939, 284 and 852 cell lines were sensitive to a dose as low as 20 ng/ml, whereas the 987 line was resistant to 2.0 micrograms/ml, even with HTX. IFN-gamma enhanced the response to TNF only of the TNF-resistant A2058 cell line, but there was no enhancement of the response to L-PAM for any line. Thus, this tetramodality combination achieved generally subadditive or additive cytoxicity in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
A2058, 91100402