EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Maximum permissible concentration of acetone cyanohydrin and cyanides in the fresh water].
PMID 7744285
Abstract
The study of general toxicological mechanism of acetone cyanhydrin and cyanides provided the data for reconsideration of their MACs in the water which is recommended at the level of 0.035 mg/l (as CN for each substance alone and both together).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A10000
Acetone cyanohydrin, 99%
C4H7NO