EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Developmental toxicity of cyproconazole, an inhibitor of fungal ergosterol biosynthesis, in the rat.
PMID 7749267
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46068
Cyproconazol, PESTANAL®, analytical standard
C15H18ClN3O