EMAIL THIS PAGE TO A FRIEND
Sangyo eiseigaku zasshi = Journal of occupational health
[Mutagenicity of herbicide chlornitrofen (CNP) and its reduction products].
PMID 7749993
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04287
Chlornitrofen, PESTANAL®, analytical standard
C12H6Cl3N1O3