EMAIL THIS PAGE TO A FRIEND
Trends in pharmacological sciences
Endothelin receptors in human coronary arteries.
PMID 7754529
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P208
PD 151,242, ≥97%, powder
C34H45N5O6