EMAIL THIS PAGE TO A FRIEND
Experimental dermatology
Quantitative screening of human stratum corneum lipids by sequential one-dimensional thin layer chromatography.
PMID 7757332
Abstract
There is a need in many studies on the epidermal permeability barrier for practical and reliable separation of all major fractions of human stratum corneum lipids. Various methods have been described including thin layer chromatography, high performance thin layer chromatography and iatroscan. However, none of these methods seems to be applicable for an inexpensive, rapid and reliable analysis of a large number of samples. Here, such a method for the separation and quantification of all major stratum corneum lipid fractions is presented. This method employs the one-dimensional separation of stratum corneum lipids using thin layer chromatography. The systems used herein are based upon various modifications of the solvents, solvent ratio and developing distance of each system. For quantification of chromatographed and charred lipids a Desaga densitometer is used. The system described here had been successfully applied to 550 samples of human stratum corneum lipids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9523
Sodium cholesteryl sulfate
C27H45NaO4S