EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Contact urticaria syndrome from sorbitan sesquioleate in a corticosteroid ointment.
PMID 7758312
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S3386
Sorbitan sesquioleate, nonionic surfactant