EMAIL THIS PAGE TO A FRIEND
Letters in applied microbiology
Biotransformation of benzyl thiocyanate by Streptomyces aureofaciens.
PMID 7765389
Abstract
Benzyl thiocyanate, a specific stimulator of chlortetracycline biosynthesis, was transformed into dibenzyl disulphide by Streptomyces aureofaciens. The disulphide stimulated chlortetracycline production to a lesser extent than did benzyl thiocyanate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
13929
Benzyl thiocyanate, ≥95.0% (GC)
C8H7NS